
2'-Fluoro-4'-methoxyacetophenone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:74457-86-6
:
Formula:C₉H₉FO₂
MDL Number:MFCD00042290
:168.17
Boiling Pt:243 °C (1013 hPa)
Melting Pt:53…54 °C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:50.0 to 54.0 deg-C
Specification Test Results
Frequently Bought Together