Order Entry
Netherlands
ContactUsLinkComponent
α,α,α-Trifluoro-m-toluonitrile ≥98.0% (by GC)
Toxic
α,α,α-Trifluoro-m-toluonitrile ≥98.0% (by GC)
Catalog # TCIAC1003-25G
Supplier:  TCI
CAS Number:  
undefined
Toxic
α,α,α-Trifluoro-m-toluonitrile ≥98.0% (by GC)
Catalog # TCIAC1003-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :171.12 g/mol
Boiling Pt:189°C
Melting Pt:19…22 °C
Density:1,2821 g/cm³ (20 °C)
Flash Pt:72°C
EINECS:206-711-4
UN:3276
ADR:6.1,III
 :GHS06
CAS Number:368-77-4
Formula:C₈H₄F₃N
MDL Number:MFCD00001807