
4-Chloro-3-nitrophenyl isocyanate ≥95.0% (by titrimetric analysis)
TCIAI0564-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to Yellow to Green powder to crystal
- Purity (Butylamine Method):min. 95.0 %
- Melting point:65.0 to 68.0 deg-C
- Solubility in Methanol:almost transparency
:
:198.57 g/mol
:65…67 °C
:Refrigerator
:2206
:6.1,III
:GHS06
:40397-96-4
:C₇H₃ClN₂O₃
:MFCD00037062