
2-(4-Chlorophenyl)ethanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1875-88-3
:
Formula:C₈H₉ClO
MDL Number:MFCD00002899
:156.61 g/mol
Boiling Pt:153°C
Density:1,189 g/cm³ (25 °C)
EINECS:217-506-4
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together