

1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:402846-78-0
:
Formula:C₉H₁₇BF₄N₂
MDL Number:MFCD03427618
:240.05 g/mol
Density:1,2 g/cm³ (20 °C)
UN:1759
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White or Colorless to Yellow powder to lump to clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together