
6-Methoxy-o-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:50868-73-0
:
Formula:C₈H₁₁NO
MDL Number:MFCD00075455
:137.18 g/mol
Boiling Pt:116°C
Melting Pt:26…29 °C
Density:1,039 g/cm³ (25 °C)
:GHS07
Specification Test Results
- Appearance:White to Brown to Dark green powder to lump
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together