

Undec-10-enoyl chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:38460-95-6
:
Formula:C₁₁H₁₉ClO
MDL Number:MFCD00000772
:202.72 g/mol
Boiling Pt:93°C
Density:0,94 g/cm³ (20 °C)
EINECS:253-951-0
UN:3265
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together