Order Entry
Netherlands
ContactUsLinkComponent
1,4-Difluoro-2-iodobenzene ≥98.0% (by GC)
Irritant
1,4-Difluoro-2-iodobenzene ≥98.0% (by GC)
Catalog # TCIAD3594-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1,4-Difluoro-2-iodobenzene ≥98.0% (by GC)
Catalog # TCIAD3594-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light red to Green clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :239.99 g/mol
Boiling Pt:183°C
Flash Pt:85°C
 :GHS07
CAS Number:2265-92-1
Formula:C₆H₃F₂I
MDL Number:MFCD03094151