Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
 
p-Phenylenediamine dihydrochloride ≥99.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
 
 
 
p-Phenylenediamine dihydrochloride ≥99.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (Neutralization titration):
    min. 99.0 %
  • Solubility in Water:
    almost transparency

 

 

Frequently Bought Together