
α,α,α,α',α',α'-Hexachloro-p-xylene ≥98.0% (by GC)
Catalog # TCIAH0064-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:108.0 to 113.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:312.84 g/mol
Merck Index:14,01303
Boiling Pt:312°C
Melting Pt:110 °C
Density:1,743 g/cm³ (20 °C)
EINECS:200-686-3
UN:1759
ADR:8,III
:GHS05
CAS Number:68-36-0
Formula:C₈H₄Cl₆
MDL Number:MFCD00000791