Order Entry
Netherlands
ContactUsLinkComponent
1,4-Bis(dimethylamino)butane ≥98.0% (by GC, titration analysis)
Flammable
Corrosive
1,4-Bis(dimethylamino)butane ≥98.0% (by GC, titration analysis)
Catalog # TCIAT1289-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
1,4-Bis(dimethylamino)butane ≥98.0% (by GC, titration analysis)
Catalog # TCIAT1289-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :144.26 g/mol
Merck Index:14,09226
Boiling Pt:100°C
Density:0,792 g/cm³ (20 °C)
Flash Pt:46°C
EINECS:203-878-5
UN:2734
ADR:8,II
 :GHS02
 :GHS05
CAS Number:111-51-3
Formula:C₈H₂₀N₂
MDL Number:MFCD00008338