


m-Phenylenediamine dihydrochloride ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:541-69-5
:
Formula:C₆H₁₀Cl₂N₂
MDL Number:MFCD00012975
:181.06 g/mol
Merck Index:14,07283
Melting Pt:277…278 °C
EINECS:208-790-0
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Light yellow to Purple powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together