
3,5-Bis(trifluoromethyl)phenol ≥96.0% (by GC)
: TCI
:349-58-6
:
:C₈H₄F₆O
:MFCD00000386
:230.11 g/mol
:88°C
:21 °C
:1,51 g/cm³ (20 °C)
:206-488-3
:GHS07
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %
Frequently Bought Together