


o-Phenylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:615-28-1
:
Formula:C₆H₁₀Cl₂N₂
MDL Number:MFCD00012966
:181.06 g/mol
Melting Pt:205°C
EINECS:210-418-7
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Absorbance:max. 0.040 (1w/v%, H2O, 420.0 nm)
- Absorbance2:max. 0.050 (1w/v%, H2O, 480.0 nm)
Specification Test Results
Frequently Bought Together