Methyl yellow
Catalog # TCIAD0231-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Orange to Brown powder to crystal
- Lambda max.:505.0 to 515.0 nm (pH 2.4)
- Absorbance:min. 0.510 (5 mg/L, pH 2.4, 505.0 to 515.0 nm)
- Transition interval (pH):(pale purplish red)2.9 to 4.0 (pale redish yellow)
Specification Test Results
About this item
:
:225.29 g/mol
Merck Index:14,03229
Boiling Pt:371 °C (1013 hPa)
Melting Pt:113…117 °C
Density:1,2233 g/cm³ (20 °C)
EINECS:200-455-7
UN:2811
ADR:6.1,III
:GHS06
:GHS08
CAS Number:60-11-7
Formula:C₁₄H₁₅N₃
MDL Number:MFCD00008308