Order Entry
Netherlands
ContactUsLinkComponent
α-Bromo-4-nitro-o-cresol ≥95.0% (by HPLC, titration analysis)
Corrosive
Irritant
α-Bromo-4-nitro-o-cresol ≥95.0% (by HPLC, titration analysis)
Catalog # TCIAH0601-5G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Irritant
α-Bromo-4-nitro-o-cresol ≥95.0% (by HPLC, titration analysis)
Catalog # TCIAH0601-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Purity (HPLC):
    min. 95.0 area%
  • Purity (Argentmetric Titration):
    min. 95.0 %

Specification Test Results

About this item

 :
 :232.03
Melting Pt:147 °C
Density:Min, 1 g/cm³ (20 °C)
EINECS:212-248-9
UN:1759
ADR:8,III
 :GHS05
 :GHS07
CAS Number:772-33-8
Formula:C₇H₆BrNO₃
MDL Number:MFCD00007340