Order Entry
Netherlands
ContactUsLinkComponent
2,4-Dimethoxybenzylamine ≥98.0% (by titrimetric analysis)
Corrosive
2,4-Dimethoxybenzylamine ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
2,4-Dimethoxybenzylamine ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together