Order Entry
Netherlands
ContactUsLinkComponent
2,3-Dimethoxybenzylamine ≥98.0% (by GC, titration analysis)
 
2,3-Dimethoxybenzylamine ≥98.0% (by GC, titration analysis)
  TCIAD4022-5G
 :  TCI
 :  
undefined
 
2,3-Dimethoxybenzylamine ≥98.0% (by GC, titration analysis)
  TCIAD4022-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :167.21
 :137°C
 :224-515-7
 :2735
 :8,III
 :GHS05
 :4393-09-3
 :C₉H₁₃NO₂
 :MFCD00052392