
1-Phenyl-2-thiourea ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:103-85-5
:
Formula:C₇H₈N₂S
MDL Number:MFCD00004933
:152.22 g/mol
Merck Index:14,07315
Boiling Pt:295 - 297 °C (1013 hPa)
Melting Pt:84…85 °C
Density:1,33 g/cm³ (20 °C)
EINECS:203-151-2
UN:2811
ADR:6.1,I
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:150.0 to 155.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together