
1-Methyl-2-thiourea ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:598-52-7
:
Formula:C₂H₆N₂S
MDL Number:MFCD00004938
Melting Pt:119 °C
EINECS:209-936-6
UN:2811
ADR:6.1,II
:90.15 g/mol
:GHS06
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Iodometric Titration):min. 98.0 %
- Melting point:120.0 to 123.0 deg-C
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together