Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,3-Diethyl-2-thiourea ≥97.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
1,3-Diethyl-2-thiourea ≥97.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Bromination Method):
    min. 97.0 %
  • Melting point:
    77.0 to 80.0 deg-C

 

Frequently Bought Together