
1,1,3,3-Tetramethyl-2-thiourea ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:2782-91-4
:
Formula:C₅H₁₂N₂S
MDL Number:MFCD00008324
:132.23 g/mol
Boiling Pt:243°C
Melting Pt:74…76 °C
EINECS:220-488-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:77.0 to 80.0 deg-C
Specification Test Results
Frequently Bought Together