
2,2'-Butyliminodiethanol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:102-79-4
:
Formula:H₃CCH₂CH₂CH₂N(CH₂CH₂OH)₂
MDL Number:MFCD00002856
:161.24 g/mol
Boiling Pt:275°C
Melting Pt:–70°C
Density:0,9681 g/cm³ (20 °C)
Flash Pt:143°C
EINECS:203-055-0
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together