
(2-Chloro-1,1-dimethylethyl)benzene ≥98.0% (by GC)
: TCI
:515-40-2
:
:C₆H₅C(CH₃)₂CH₂Cl
:MFCD00000940
:168.67 g/mol
:14,06459
:222°C
:1,0379 g/cm³ (20 °C)
:96°C
:208-197-7
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together