N-Ethylbenzylamine ≥97.0% (by GC, titration analysis)
: TCI
:14321-27-8
:
:C₆H₅CH₂NHCH₂CH₃
:MFCD00009031
:135.21 g/mol
:199°C
:0,909 g/cm³ (20 °C)
:69°C
:238-265-1
:2735
:8,III
:GHS05
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Frequently Bought Together