
1-(2-Methylphenyl)-2-thiourea ≥98.0% (by HPLC)
TCIAT0657-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Solubility in Methanol:very faint turbidity
:
:166.25 g/mol
:295 °C (1013 hPa)
:160 °C
:210-395-3
:2811
:6.1,III
:GHS06
:614-78-8
:(C₆H₄(CH₃))NC(S)NH₂
:MFCD00041162