(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Supplier: TCI
Certificates
About this item
CAS Number:76189-56-5
:
Formula:C₄₄H₃₂P₂
MDL Number:MFCD00010805
:622.69 g/mol
Merck Index:13,01223
Melting Pt:241…242 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity:min. 98.0 area%
- Optical purity:min. 98.0 ee%
Specification Test Results
Related Information
Frequently Bought Together