cis-4-Hydroxy-L-(-)-proline ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:618-27-9
:
Formula:C₅H₉NO₃
MDL Number:MFCD00064319
:131.13 g/mol
Merck Index:14,04840
Melting Pt:241°C
EINECS:210-542-1
Specification Test Results
- Appearance:White to Light yellow powder to crystaline
- Purity (Nonaqueous Titration):min. 97.0 %
- Specific rotation [a]20/D:-56.0 to -60.0 deg (C=2, H2O)
Specification Test Results
Frequently Bought Together