Order Entry
Netherlands
ContactUsLinkComponent
D-(+)-Proline ≥98.0% (by titrimetric analysis)
D-(+)-Proline ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
D-(+)-Proline ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +83.0 to +87.0 deg (C=4, H2O)
  • Solubility in Water:
    within almost transparency

Specification Test Results

Frequently Bought Together