


Ethyl lactate ≥98.0% (by GC)
: TCI
:97-64-3
:687-47-8
:C₅H₁₀O₃
:MFCD00065359
:118.13 g/mol
:14,03817
:154°C
:–25°C
:1,042 g/cm³ (20 °C)
:53°C
:Ambient
:202-598-0
:1192
:3,III
:GHS02
:GHS05
:GHS07
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together