
2,6-Dimethyl-4-pyrone ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1004-36-0
:
Formula:C₇H₈O₂
MDL Number:MFCD00006579
Boiling Pt:248 °C (1013 hPa)
Melting Pt:130…131 °C
Density:1,17 g/cm³ (25 °C)
:124.14 g/mol
EINECS:213-719-1
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (GC):min. 99.0 %
- Melting point:132.0 to 136.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together