
1,8-Naphthalenedicarboxylic anhydride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:81-84-5
:
Formula:C₁₂H₆O₃
MDL Number:MFCD00006925
Boiling Pt:425 °C (1013 hPa)
Melting Pt:274°C
:198.18 g/mol
EINECS:201-380-2
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Methanolysis Method):min. 98.0 %
Specification Test Results
Frequently Bought Together