Cetrimonium chloride ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:112-02-7
:
Formula:C₁₉H₄₂ClN
MDL Number:MFCD00011773
:320 g/mol
Melting Pt:232…233 °C
Density:0,9 g/cm³ (20 °C)
EINECS:203-928-6
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 95.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
- Solubility in Water:almost transparency
- Water:max. 5.0 %
Specification Test Results
Frequently Bought Together