
1,3-Bis(diphenylphosphino)propane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6737-42-4
:
Formula:C₂₇H₂₆P₂
MDL Number:MFCD00003050
Melting Pt:60…63 °C
:412.45 g/mol
EINECS:229-791-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Light yellow powder to crystaline
- Purity (GC):min. 98.0 %
- Purity (Iodometric Titration):min. 96.0 %
- Melting point:58.0 to 63.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together