Tetrachloro-p-benzoquinone ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:118-75-2
:
Formula:C₆Cl₄O₂
MDL Number:MFCD00001594
:245.88 g/mol
Merck Index:14,02078
Boiling Pt:270 °C (1013 hPa)
Melting Pt:297°C
Density:1,97 g/cm³ (20 °C)
Flash Pt:≥ 100 °C
EINECS:204-274-4
UN:3077
ADR:9,III
:GHS05
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystaline
- Purity (GC):min. 98.0 %
- Purity (Iodometric Titration):min. 98.0 %
- Solubility in hot Toluene:almost transparency
Specification Test Results
Frequently Bought Together