Order Entry
Netherlands
ContactUsLinkComponent
2-tert-Butyl-p-cresol ≥98.0% (by GC)
Corrosive
2-tert-Butyl-p-cresol ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Corrosive
2-tert-Butyl-p-cresol ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    51.0 to 54.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Frequently Bought Together