



6-tert-Butyl-m-cresol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:88-60-8
:
Formula:C₁₁H₁₆O
MDL Number:MFCD00002308
:164.25
Boiling Pt:244°C
Melting Pt:20…23 °C
Density:0,964 g/cm³ (20 °C)
EINECS:201-842-3
UN:2430
ADR:8,III
:GHS05
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together