
Dimethyl-5-aminoisophthalate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA1586-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:178.0 to 182.0 deg-C
Specification Test Results
About this item
:
:209.2 g/mol
EINECS:202-743-8
:GHS07
CAS Number:99-27-4
Formula:C₁₀H₁₁NO₄
MDL Number:MFCD00008435