Order Entry
Netherlands
ContactUsLinkComponent
β-(N-Morpholino)ethanesulphonic acid (MES) ≥99.0% (by titrimetric analysis)
β-(N-Morpholino)ethanesulphonic acid (MES) ≥99.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
β-(N-Morpholino)ethanesulphonic acid (MES) ≥99.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (Neutralization titration):
    min. 99.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together