Order Entry
Netherlands
ContactUsLinkComponent
1,6-Dimethoxyhexane ≥97.0% (by GC)
1,6-Dimethoxyhexane ≥97.0% (by GC)
Catalog # TCIAD2858-5G
Supplier:  TCI
CAS Number:  
1,6-Dimethoxyhexane ≥97.0% (by GC)
Catalog # TCIAD2858-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
Flash Pt:69°C
 :146.23 g/mol
CAS Number:13179-98-1
Formula:C₈H₁₈O₂
MDL Number:MFCD00142569