Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
 
3-(Dimethylamino)-1-propanol ≥98.0% (by GC, titration analysis)
  TCIAD0661-25ML
 :  TCI
 :  
undefined
 
 
 
3-(Dimethylamino)-1-propanol ≥98.0% (by GC, titration analysis)
  TCIAD0661-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

 

 

 :
 :103.16 g/mol
 :164°C
 :–35 °C
 :0,8878 g/cm³ (20 °C)
 :36°C
 :221-659-2
 :2920
 :8,II
 :GHS02
 :GHS05
 :GHS07
 :3179-63-3
 :C₅H₁₃NO
 :MFCD00002945