Order Entry
Netherlands
ContactUsLinkComponent
Sodium thiosulphate concentrate for dilution to 0.01 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Sodium thiosulphate concentrate for dilution to 0.01 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 84598.600
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium thiosulphate concentrate for dilution to 0.01 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 84598.600
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    60 mL
  • Seq:
    2385

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.00998 - 0.01020 mol/l

Specification Test Results

About this item

 :
 :158.11 g/mol
 :Dose for preparation of 1 litre of solution.
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499