
Tetramethylammonium iodide
Supplier: TCI
Certificates
About this item
CAS Number:75-58-1
:
Formula:C₄H₁₂IN
MDL Number:MFCD00011629
Merck Index:14,09225
Melting Pt:300 °C
Density:1,84 g/cm³ (20 °C)
EINECS:200-881-3
ADR:6.1,III
:201.05 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Potassium iodate Method):min. 98.0 %
- Drying loss:max. 1.0 %
- pH:4.0 to 7.0
Specification Test Results
Related Information
Frequently Bought Together