
1,1-Cyclopropanedicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:598-10-7
:
Formula:C₅H₆O₄
MDL Number:MFCD00013727
:130.1 g/mol
Boiling Pt:371 °C (1013 hPa)
Melting Pt:134…138 °C
EINECS:209-917-2
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:137.0 to 143.0 deg-C
- Solubility in Water:very faint turbidity
Specification Test Results
Frequently Bought Together