


Cyclopropanecarbonyl chloride ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4023-34-1
:
Formula:C₄H₅ClO
MDL Number:MFCD00001277
:104.54 g/mol
Boiling Pt:119°C
Density:1,15 g/cm³ (20 °C)
Flash Pt:23°C
EINECS:223-684-4
UN:2920
ADR:8,II
:GHS02
:GHS05
:GHS06
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Purity (Argentmetric Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together