Order Entry
Netherlands
ContactUsLinkComponent
1,5-Diphenylcarbazide ≥98.0% (by HPLC)
1,5-Diphenylcarbazide ≥98.0% (by HPLC)
Catalog # TCIAD1513-25G
Supplier:  TCI
CAS Number:  
undefined
1,5-Diphenylcarbazide ≥98.0% (by HPLC)
Catalog # TCIAD1513-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Solubility in Acetone:
    almost transparency
  • Suitability for chrome-analysis:
    Abs min.0.42 (near 540nm)in the presence of Dichromate (1 ppm)

Specification Test Results

About this item

 :
 :242.28 g/mol
Merck Index:14,03321
Boiling Pt:454 °C (1013 hPa)
Melting Pt:170…172 °C
Density:1,31 g/cm³ (20 °C)
Storage Temperature:Freezer
EINECS:205-403-7
 :GHS07
CAS Number:140-22-7
Formula:C₁₃H₁₄N₄O
MDL Number:MFCD00003013