
2,2'-Dinitrobiphenyl ≥99.0% (by GC)
: TCI
:2436-96-6
:
:C₁₂H₈N₂O₄
:MFCD00007127
:244.21 g/mol
:305 °C (1013 hPa)
:123…125 °C
:1,45 g/cm³ (20 °C)
:Ambient
:219-435-4
:6.1,II
:GHS07
- Purity (GC):min. 99.0 %
- Melting point:124.0 to 127.0 deg-C
Frequently Bought Together