Order Entry
Netherlands
ContactUsLinkComponent
 
 
2-Mesitylenesulphonyl chloride ≥99.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
2-Mesitylenesulphonyl chloride ≥99.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White powder to crystal
  • Purity (Neutralization titration):
    min. 99.0 %
  • Melting point:
    54.0 to 58.0 deg-C
  • Solubility in Toluene:
    almost transparency

 

Frequently Bought Together