2,2'-Dimethylbiphenyl ≥95.0% (by GC)
: TCI
:605-39-0
:
:C₁₄H₁₄
:MFCD00048075
:182.27 g/mol
:259°C
:18 °C
:0,993 g/cm³ (20 °C)
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Frequently Bought Together