

3,5-Dimethylbenzoyl chloride ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6613-44-1
:
Formula:C₉H₉ClO
MDL Number:MFCD02313742
:168.62 g/mol
Boiling Pt:127°C
EINECS:413-010-9
UN:3265
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
- Purity (Argentmetric Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together